tert-Butyl vinyl ether 98% stabilised
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with N,N-diethylaniline ∼ 0.1%
H30770-06
H30770-14
AAH30770-06EA
75.6
USD
AAH30770-06
AAH30770-14
tert-Butyl vinyl ether 98% stabilised
tert-Butyl vinyl ether
Stab. with ca 0.1% N,N-diethylaniline
Purity: 98%
CAS No.: 926-02-3
Molecular Formula: C6H12O
Formula Weight: 100.16
Storage Temperature: 2°C to 8°C
Flash Point: -17°(1°F)
Refractive Index: 1.3990
Odor: Characteristic
Boiling Point: 75-76°
UN No.: 3271
MDL No.: MFCD00048246
Formula:
C₆H₁₂O Boiling Pt: 75…76 °C Density: 0.762 Flash Pt: –17 °C (1 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00048246 CAS Number: 926-02-3 EINECS: 618-860-6 UN: 3271 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...