3-Bromo-1-propylboronic acid pinacol ester 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H30646-03
H30646-06
AAH30646-03EA
51.4
USD
AAH30646-03
AAH30646-06
3-Bromo-1-propylboronic acid pinacol ester 98%
3-Bromo-1-propylboronic acid pinacol ester
Purity: 98%
CAS No.: 124215-44-7
Molecular Formula: C9H18BBrO2
Formula Weight: 248.96
Boiling Point: 61-64°/0.35mm
MDL No.: MFCD10567053
Formula:
C₉H₁₈BBrO₂ Boiling Pt: 61…64 °C/0.35 mm Density: 1.177 Storage Temperature: Ambient |
MDL Number:
MFCD10567053 CAS Number: 124215-44-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...