Thieno[3,2-b]thiophene 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,4-Dithiapentalene

H30637-03 H30637-06
AAH30637-03EA 151 USD
AAH30637-03 AAH30637-06
Thieno[3,2-b]thiophene 97%
Thieno[3,2-b]thiophene

Purity: 97% CAS No.: 251-41-2 Molecular Formula: C6H4S2 Formula Weight: 140.23 Melting Point: 54-58° Boiling Point: 71°/1mm MDL No.: MFCD00179479

Formula: C₆H₄S₂
MW: 140.23 g/mol
Boiling Pt: 71 °C/1 mm
Melting Pt: 54…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00179479
CAS Number: 251-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR