2-Amino-6-fluoropyridine 97%

Supplier: Thermo Scientific Chemicals
Danger

H30629-03 H30629-06
AAH30629-03EA 104 USD
AAH30629-03 AAH30629-06
2-Amino-6-fluoropyridine 97%
2-Amino-6-fluoropyridine

Purity: 97% CAS No.: 1597-32-6 Molecular Formula: C5H5FN2 Formula Weight: 112.11 Odor: Characteristic Melting Point: 58-62° UN No.: 2811 MDL No.: MFCD04114200

Formula: C₅H₅FN₂
MW: 112.11 g/mol
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD04114200
CAS Number: 1597-32-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR