3-Bromo-4-fluorobenzyl alcohol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3-Bromo-4-fluorophenyl)methanol

H30604-06 H30604-14
AAH30604-06EA 31.9 USD
AAH30604-06 AAH30604-14
3-Bromo-4-fluorobenzyl alcohol 97%
3-Bromo-4-fluorobenzyl alcohol

Purity: 97% CAS No.: 77771-03-0 Molecular Formula: C7H6BrFO Formula Weight: 205.03 Boiling Point: 80°/0.5mm MDL No.: MFCD00143093

Formula: C₇H₆BrFO
MW: 205.03 g/mol
Boiling Pt: 80 °C/0.5 mm
Storage Temperature: Ambient
MDL Number: MFCD00143093
CAS Number: 77771-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR