2-Bromo-6-methoxypyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H30593-06 H30593-14
AAH30593-06EA 44.9 USD
AAH30593-06 AAH30593-14
2-Bromo-6-methoxypyridine 97%
2-Bromo-6-methoxypyridine

Purity: 97% CAS No.: 40473-07-2 Molecular Formula: C6H6BrNO Formula Weight: 188.02 Flash Point: 104°(219°F) Refractive Index: 1.5600 Boiling Point: 206° MDL No.: MFCD00088345

Formula: C₆H₆BrNO
MW: 188.02 g/mol
Boiling Pt: 206 °C
Density: 1.530
Flash Pt: 104 °C(219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00088345
CAS Number: 40473-07-2
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR