2-(3-Chlorophenyl)ethylboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
H30464-06
H30464-03
AAH30464-06EA
116
USD
AAH30464-06
AAH30464-03
2-(3-Chlorophenyl)ethylboronic acid pinacol ester 97%
2-(3-Chlorophenyl)ethylboronic acid pinacol ester
Purity: 97%
Molecular Formula: C14H20BClO2
Formula Weight: 266.58
Refractive Index: 1.4965
MDL No.: MFCD12546188
Formula:
C₁₄H₂₀BClO₂ MW: 266.58 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12546188 CAS Number: 1315277-53-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...