2,6-Dichloropyridine-N-oxide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,6-Dichloropyridine 1-oxide

H30427-03 H30427-06
AAH30427-03EA 31.8 USD
AAH30427-03 AAH30427-06
2,6-Dichloropyridine-N-oxide 98%
2,6-Dichloropyridine N-oxide

Purity: 98% CAS No.: 2587-00-0 Molecular Formula: C5H3Cl2NO Formula Weight: 163.99 Melting Point: 136-140° UN No.: 2811 MDL No.: MFCD00130246

Formula: C₅H₃Cl₂NO
MW: 163.99 g/mol
Melting Pt: 136…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00130246
CAS Number: 2587-00-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR