1,4-Bis(phenylethynyl)benzene 97%
Supplier: Thermo Scientific Chemicals
H30395-03
H30395-14
H30395-06
AAH30395-03EA
47.2
USD
AAH30395-03
AAH30395-14
AAH30395-06
1,4-Bis(phenylethynyl)benzene 97%
1,4-Bis(phenylethynyl)benzene
Purity: 97%
CAS No.: 1849-27-0
Molecular Formula: C22H14
Formula Weight: 278.36
Melting Point: 176-178°
MDL No.: MFCD00160829
Formula:
C₂₂H₁₄ Melting Pt: 176…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00160829 CAS Number: 1849-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...