2-Acetyl-4-chlorothiophene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(4-Chlorothiophen-2-yl)ethanone, 1-(4-Chloro-2-thienyl)-1-ethanone
H30384-06
H30384-14
AAH30384-03EA
39.34
USD
AAH30384-03
AAH30384-06
AAH30384-14
2-Acetyl-4-chlorothiophene 98+%
2-Acetyl-4-chlorothiophene
Purity: 98+%
CAS No.: 34730-20-6
Molecular Formula: C6H5ClOS
Formula Weight: 160.62
UN No.: 2810
MDL No.: MFCD00082791
Formula:
C₆H₅ClOS MW: 160.62 g/mol Density: 1.334 Storage Temperature: Ambient |
MDL Number:
MFCD00082791 CAS Number: 34730-20-6 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...