Monomethyl Suberate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 8-Methoxy-8-oxooctanoic acid, Suberic Acid Monomethyl Ester, Methyl Hydrogen Suberate, Octanedioic Acid Monomethyl Ester

H30325-03 H30325-14 H30325-06
AAH30325-03EA 60.2 USD
AAH30325-03 AAH30325-14 AAH30325-06
Monomethyl Suberate 98%
Monomethyl suberate

Purity: 98% CAS No.: 3946-32-5 Molecular Formula: C9H16O4 Formula Weight: 188.22 Melting Point: 17-19° Boiling Point: 185-186°/18mm MDL No.: MFCD00004427

Formula: C₉H₁₆O₄
MW: 188.22 g/mol
Boiling Pt: 185…186 °C/18 mm
Melting Pt: 17…19 °C
Density: 1.04
Storage Temperature: Ambient
MDL Number: MFCD00004427
CAS Number: 3946-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR