1H-Pyrrolo[2,3-b]pyridine-5-carbonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

H30237-03 H30237-MD
AAH30237-03EA 98.3 USD
AAH30237-03 AAH30237-MD
1H-Pyrrolo[2,3-b]pyridine-5-carbonitrile 97%
1H-Pyrrolo[2,3-b]pyridine-5-carbonitrile

Purity: 97% CAS No.: 517918-95-5 Molecular Formula: C8H5N3 Formula Weight: 143.15 Melting Point: 226-230° MDL No.: MFCD06659684

Formula: C₈H₅N₃
MW: 143.15 g/mol
Melting Pt: 226…230 °C
Storage Temperature: Ambient
MDL Number: MFCD06659684
CAS Number: 517918-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR