5-Oxazolecarboxylic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Oxazole-5-carboxylic acid
H30179-03
H30179-06
AAH30179-03EA
61
USD
AAH30179-03
AAH30179-06
5-Oxazolecarboxylic acid 98+%
5-Oxazolecarboxylic acid
Purity: 98+%
CAS No.: 118994-90-4
Molecular Formula: C4H3NO3
Formula Weight: 113.07
Melting Point: ca 245° dec.
MDL No.: MFCD04114931
Formula:
C₄H₃NO₃ MW: 113.07 g/mol Melting Pt: ca 245 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD04114931 CAS Number: 118994-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...