3-Chloro-5-fluoropyridine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H30170-03
H30170-06
AAH30170-03EA
133
USD
AAH30170-03
AAH30170-06
3-Chloro-5-fluoropyridine 98%
3-Chloro-5-fluoropyridine
Purity: 98%
CAS No.: 514797-99-0
Molecular Formula: C5H3ClFN
Formula Weight: 131.54
Refractive Index: 1.5015
Melting Point: 25-27°
UN No.: 1325
MDL No.: MFCD05663707
Formula:
C₅H₃ClFN Melting Pt: 25…27 °C Flash Pt: 41 °C (106 °F) Storage Temperature: Ambient |
MDL Number:
MFCD05663707 CAS Number: 514797-99-0 UN: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...