4-Benzyloxy-1-bromo-2-fluorobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

H30053-06 H30053-14
AAH30053-06EA 78.9 USD
AAH30053-06 AAH30053-14
4-Benzyloxy-1-bromo-2-fluorobenzene 98%
4-Benzyloxy-1-bromo-2-fluorobenzene

Purity: 98% CAS No.: 185346-79-6 Molecular Formula: C13H10BrFO Formula Weight: 281.13 Melting Point: 33-35° MDL No.: MFCD09801041

Formula: C₁₃H₁₀BrFO
Melting Pt: 33…35 °C
Storage Temperature: Ambient
MDL Number: MFCD09801041
CAS Number: 185346-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR