Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, (H₂O 1-5%) 97% may cont. 1-5% water

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Kobayashi's Reagent, NaBArF24

H30014-14 H30014-03 H30014-06
AAH30014-14EA 1784 USD
AAH30014-14 AAH30014-03 AAH30014-06
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, (H₂O 1-5%) 97% may cont. 1-5% water
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate

Purity: 97% CAS No.: 79060-88-1 Molecular Formula: C32H12BF24Na Formula Weight: 886.20 Melting Point: 310° dec. MDL No.: MFCD00043323

Formula: C₃₂H₁₂BF₂₄Na
MW: 886.20 g/mol
Melting Pt: 310 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00043323
CAS Number: 79060-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR