Benzofurazan-5-boronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzofurazan, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[c][1,2,5]oxadiazole

H29358-03 H29358-06 H29358-MD
AAH29358-03EA 179 USD
AAH29358-03 AAH29358-06 AAH29358-MD
Benzofurazan-5-boronic acid pinacol ester 97%
Benzofurazan-5-boronic acid pinacol ester

Purity: 97% CAS No.: 1073355-14-2 Molecular Formula: C12H15BN2O3 Formula Weight: 246.07 Melting Point: 87-88° MDL No.: MFCD09842717

Formula: C₁₂H₁₅BN₂O₃
MW: 246.07 g/mol
Melting Pt: 87…88 °C
Storage Temperature: Ambient
MDL Number: MFCD09842717
CAS Number: 1073355-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR