3-Carboxy-5-nitrophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Carboxy-5-nitrobenzeneboronic acid, 3-Borono-5-nitrobenzoic acid

H29348-03 H29348-06
AAH29348-03EA 61.7 USD
AAH29348-03 AAH29348-06
3-Carboxy-5-nitrophenylboronic acid 97%
3-Carboxy-5-nitrophenylboronic acid

Purity: 97% CAS No.: 101084-81-5 Molecular Formula: C7H6BNO6 Formula Weight: 210.94 MDL No.: MFCD00757433

Formula: C₇H₆BNO₆
MW: 210.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00757433
CAS Number: 101084-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR