5-Formyl-2-thiopheneboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Formylthiophen-2-boronic acid, 5-Formylthiophene-2-boronic acid

H29289-03 H29289-06
AAH29289-03EA 70.5 USD
AAH29289-03 AAH29289-06
5-Formyl-2-thiopheneboronic acid 97%
5-Formyl-2-thiopheneboronic acid

Purity: 97% CAS No.: 4347-33-5 Molecular Formula: C5H5BO3S Formula Weight: 155.97 Storage Temperature: 2°C to 8°C Melting Point: ca 124° dec. MDL No.: MFCD02093666

Formula: C₅H₅BO₃S
MW: 155.97 g/mol
Melting Pt: ca 124 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD02093666
CAS Number: 4347-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR