2,6-Dichlorobenzylamine 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H29274-03
H29274-14
AAH29274-03EA
17.5
USD
AAH29274-03
AAH29274-14
2,6-Dichlorobenzylamine 97+%
2,6-Dichlorobenzylamine
Purity: 97+%
CAS No.: 6575-27-5
Molecular Formula: C7H7Cl2N
Formula Weight: 176.05
Odor: Amine-like
UN No.: 2735
MDL No.: MFCD00047928
Formula:
C₇H₇Cl₂N Storage Temperature: Ambient |
MDL Number:
MFCD00047928 CAS Number: 6575-27-5 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...