(±)-1-Boc-3-aminopyrrolidine 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Aminopyrrolidine, N1-BOC protected, 1-Boc-3-Aminopyrrolidine
H29231-03
H29231-06
AAH29231-03EA
63
USD
AAH29231-03
AAH29231-06
AAH29231-MD
(±)-1-Boc-3-aminopyrrolidine 96%
(±)-1-Boc-3-aminopyrrolidine
Purity: 96%
CAS No.: 186550-13-0
Molecular Formula: C9H18N2O2
Formula Weight: 186.25
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4710
UN No.: 2735
MDL No.: MFCD01861220
Formula:
C₉H₁₈N₂O₂ MW: 186.25 g/mol Density: 1.022 Storage Temperature: Ambient |
MDL Number:
MFCD01861220 CAS Number: 186550-13-0 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...