4'-Bromo-2'-hydroxy-5'-methylacetophenone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H29125-03
H29125-06
AAH29125-03EA
23.3
USD
AAH29125-03
AAH29125-06
4'-Bromo-2'-hydroxy-5'-methylacetophenone 97%
4'-Bromo-2'-hydroxy-5'-methylacetophenone
Purity: 97%
CAS No.: 50342-17-1
Molecular Formula: C9H9BrO2
Formula Weight: 229.07
Melting Point: 86-89°
MDL No.: MFCD08273792
Formula:
C₉H₉BrO₂ Melting Pt: 86…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD08273792 CAS Number: 50342-17-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...