2-Aminoacetophenone hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Phenacylamine hydrochloride
H29117-03
H29117-09
AAH29117-18EA
710.96
USD
AAH29117-18
AAH29117-03
AAH29117-09
2-Aminoacetophenone hydrochloride 97%
2-Aminoacetophenone hydrochloride
Purity: 98%
CAS No.: 5468-37-1
Molecular Formula: C8H9NO.HCl
Formula Weight: 171.63
Odor: Odorless
Melting Point: ca 194° dec.
MDL No.: MFCD00012873
Formula:
C₈H₁₀ClNO MW: 171.63 g/mol Melting Pt: ca 194 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012873 CAS Number: 5468-37-1 EINECS: 226-787-2 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...