4-Methyl-1,2-cyclohexene oxide cis- and trans-mixture 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H29100-03
H29100-14
H29100-06
AAH29100-03EA
42.8
USD
AAH29100-03
AAH29100-14
AAH29100-06
4-Methyl-1,2-cyclohexene oxide cis- and trans-mixture 97%
4-Methyl-1,2-cyclohexene oxide
Purity: 97%
CAS No.: 36099-51-1
Molecular Formula: C7H12O
Formula Weight: 112.17
Refractive Index: 1.4470
MDL No.: MFCD09742280
Formula:
C₇H₁₂O Storage Temperature: Ambient |
MDL Number:
MFCD09742280 CAS Number: 36099-51-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...