Vinyl decanoate 95% stabilized
Supplier: Thermo Scientific Chemicals
H29081-14
H29081-06
AAH29081-14EA
90.3
USD
AAH29081-14
AAH29081-06
Vinyl decanoate 95% stabilized
Vinyl decanoate
Purity: 95%
CAS No.: 4704-31-8
Molecular Formula: C12H22O
Formula Weight: 198.30
Flash Point: 104°(219°F)
Refractive Index: 1.435
Boiling Point: 119-120°/10mm
MDL No.: MFCD00027349
Formula:
C₁₂H₂₂O₂ Boiling Pt: 119…120 °C/10 mm Density: 0.886 Flash Pt: 104 °C(219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00027349 CAS Number: 4704-31-8 EINECS: 225-185-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...