Cycloheptene 96% stabilized

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 0.1%

H29062-06 H29062-14
AAH29062-06EA 112 USD
AAH29062-06 AAH29062-14
Cycloheptene 96% stabilized
Cycloheptene

Purity: 96% CAS No.: 628-92-2 Molecular Formula: C7H12 Formula Weight: 96.17 Flash Point: -7°(19°F) Refractive Index: 1.458 Boiling Point: 112-115° UN No.: 2242 MDL No.: MFCD00004156

Formula: C₇H₁₂
MW: 96.17 g/mol
Boiling Pt: 112…115 °C
Melting Pt: -56 °C
Density: 0.824 g/cm³ (20 °C)
Flash Pt: –7 °C (19 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004156
CAS Number: 628-92-2
EINECS: 211-060-4
UN: 2242
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR