N-Fmoc-D-leucine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-D-Leu-OH
H29041-14
H29041-03
H29041-06
AAH29041-14EA
275
USD
AAH29041-14
AAH29041-03
AAH29041-06
N-Fmoc-D-leucine 98%
N-Fmoc-D-leucine
Purity: 98%
CAS No.: 114360-54-2
Molecular Formula: C21H23NO4
Formula Weight: 353.42
Storage Temperature: 2°C to 8°C
Melting Point: 151-154°
Optical Rotation: +25° (c=1 in DMF)
MDL No.: MFCD00062957
Formula:
C₂₁H₂₃NO₄ MW: 353.4 g/mol Melting Pt: 151…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD00062957 CAS Number: 114360-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...