2-(Boc-amino)benzeneboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Boc-amino)phenylboronic Acid Pinacol Ester
H29020-03
H29020-06
AAH29020-03EA
64.8
USD
AAH29020-03
AAH29020-06
2-(Boc-amino)benzeneboronic acid pinacol ester 97%
2-(Boc-amino)benzeneboronic acid pinacol ester
Purity: 97%
CAS No.: 159624-15-4
Molecular Formula: C17H26BNO4
Formula Weight: 319.21
Melting Point: 98-100°
MDL No.: MFCD03411943
Formula:
C₁₇H₂₆BNO₄ MW: 319.21 g/mol Melting Pt: 98…100 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411943 CAS Number: 159624-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...