(S)-Piperazine-2-carboxylic acid dihydrochloride ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2S)-piperazine-2-carboxylic acid dihydrochloride

H28972-03 H28972-06
AAH28972-03EA 54.2 USD
AAH28972-03 AAH28972-MD AAH28972-06
(S)-Piperazine-2-carboxylic acid dihydrochloride ≥98%
(S)-Piperazine-2-carboxylic acid dihydrochloride

Purity: 98+% CAS No.: 158663-69-5 Molecular Formula: C5H10N2O2.2HCl Formula Weight: 203.07 Melting Point: ca 280° dec. Optical Rotation: -5° (c=1 in water) MDL No.: MFCD01632104

Formula: C₅H₁₂Cl₂N₂O₂
Melting Pt: ∼ 280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01632104
CAS Number: 158663-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR