4-(Methoxycarbonyl)phenylboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Methoxycarbonyl)benzeneboronic acid pinacol ester, Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
H28942-03
H28942-06
AAH28942-03EA
28.7
USD
AAH28942-03
AAH28942-06
4-(Methoxycarbonyl)phenylboronic acid pinacol ester 97%
4-(Methoxycarbonyl)phenylboronic acid pinacol ester
Purity: 97%
CAS No.: 171364-80-0
Molecular Formula: C14H19BO4
Formula Weight: 262.11
Melting Point: 77-81°
MDL No.: MFCD02179438
Formula:
C₁₄H₁₉BO₄ MW: 262.11 g/mol Melting Pt: 77…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179438 CAS Number: 171364-80-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...