tert-Butyl-3-hydroxyazetidine-1-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Boc-3-hydroxyazetidine, tert-Butyl 3-hydroxyazetidine-1-carboxylate, 1-BOC-3-Hydroxyazetidine, tert-butyl 3-hydroxyazetidine-1-carboxylate

H28902-03 H28902-06
AAH28902-03EA 52.7 USD
AAH28902-03 AAH28902-06 AAH28902-MD
tert-Butyl-3-hydroxyazetidine-1-carboxylate 97%
tert-Butyl-3-hydroxyazetidine-1-carboxylate

Purity: 97% CAS No.: 141699-55-0 Molecular Formula: C8H15NO3 Formula Weight: 173.21 Melting Point: 40-47° MDL No.: MFCD04115305

Formula: C₈H₁₅NO₃
MW: 173.21 g/mol
Melting Pt: 40…47 °C
Storage Temperature: Ambient
MDL Number: MFCD04115305
CAS Number: 141699-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR