N-Boc-azetidine-3-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Boc-azetidine-3-carboxylic acid, N-Boc-Azetidine-3-carboxylic acid, 1-N-Boc-3-Azetidinecarboxylic acid, 1-Boc-azetidine-3-carboxylic Acid
H28817-06
H28817-03
AAH28817-06EA
749
USD
AAH28817-06
AAH28817-MD
AAH28817-03
N-Boc-azetidine-3-carboxylic acid 97%
N-Boc-azetidine-3-carboxylic acid
Purity: 97%
CAS No.: 142253-55-2
Molecular Formula: C9H15NO4
Formula Weight: 201.22
Melting Point: 98-105°
MDL No.: MFCD01860897
Formula:
C₉H₁₅NO₄ MW: 201.22 g/mol Melting Pt: 98…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD01860897 CAS Number: 142253-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...