(S)-(-)-4-Amino-2-hydroxybutyric acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-4-Amino-2-hydroxybutanoic acid

H28796-06 H28796-14
AAH28796-06EA 64.8 USD
AAH28796-06 AAH28796-14
(S)-(-)-4-Amino-2-hydroxybutyric acid 98%
(S)-(-)-4-Amino-2-hydroxybutyric Acid

Purity: 98% CAS No.: 40371-51-5 Molecular Formula: C4H9NO3 Formula Weight: 119.12 MDL No.: MFCD00674110

Formula: C₄H₉NO₃
MW: 119.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00674110
CAS Number: 40371-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR