L-Serine benzyl ester hydrochloride 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-Ser-OBzl·HCl, H-Ser-OBzl.HCl
H28790-03
H28790-06
H28790-14
AAH28790-03EA
46.5
USD
AAH28790-03
AAH28790-06
AAH28790-14
L-Serine benzyl ester hydrochloride 98%
L-Serine benzyl ester hydrochloride
Purity: 98%
CAS No.: 60022-62-0
Molecular Formula: C10H13NO3.HCl
Formula Weight: 231.68
Storage Temperature: 2°C to 8°C
Melting Point: ca 175° dec.
Optical Rotation: -12° (c=4 in water)
MDL No.: MFCD00038955
Formula:
C₁₀H₁₃NO₃·HCl MW: 231.6 g/mol Melting Pt: ca 175 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00038955 CAS Number: 60022-62-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...