2-Fluoro-4-(trifluoromethyl)phenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Fluoro-4-(trifluoromethyl)benzeneboronic acid

H28788-06 H28788-03
AAH28788-06EA 356 USD
AAH28788-06 AAH28788-03
2-Fluoro-4-(trifluoromethyl)phenylboronic acid 97%
2-Fluoro-4-(trifluoromethyl)phenylboronic acid

Purity: 97% CAS No.: 503309-11-3 Molecular Formula: C7H5BF4O2 Formula Weight: 207.92 Melting Point: 113-115° MDL No.: MFCD03094995

Formula: C₇H₅BF₄O₂
Melting Pt: 113…115 °C
Storage Temperature: Ambient
MDL Number: MFCD03094995
CAS Number: 503309-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR