5-(Trifluoromethyl)indole 98%

Supplier: Thermo Scientific Chemicals
Warning

H28782-MC H28782-ME
AAH28782-MCEA 94.2 USD
AAH28782-MC AAH28782-ME
5-(Trifluoromethyl)indole 98%
5-(Trifluoromethyl)indole

Purity: 98% CAS No.: 100846-24-0 Molecular Formula: C9H6F3N Formula Weight: 185.15 Melting Point: 68-71° MDL No.: MFCD03095341

Formula: C₉H₆F₃N
MW: 185.15 g/mol
Melting Pt: 68…71 °C
Storage Temperature: Ambient
MDL Number: MFCD03095341
CAS Number: 100846-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR