3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4,4,5,5-Tetramethyl-2-[3-(4-Methyl-1-piperazinylmethyl)phenyl]-1,3,2-dioxaborolane, 1-Methyl-4-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)piperazine, 3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid Pinacol ester, 1-Methyl-4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperazine
H28762-06
H28762-03
AAH28762-06EA
339
USD
AAH28762-06
AAH28762-03
3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester 97%
3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester
Purity: 97%
CAS No.: 883738-27-0
Molecular Formula: C18H29BN2O2
Formula Weight: 316.25
MDL No.: MFCD09266181
Formula:
C₁₈H₂₉BN₂O₂ MW: 316.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09266181 CAS Number: 883738-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...