Methyl-3-iodo-4-methylbenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H28734-06
H28734-14
AAH28734-06EA
61.5
USD
AAH28734-06
AAH28734-14
Methyl-3-iodo-4-methylbenzoate 98%
Methyl 3-iodo-4-methylbenzoate
Purity: 98%
CAS No.: 90347-66-3
Molecular Formula: C9H9IO2
Formula Weight: 276.07
Refractive Index: 1.5975
Formula:
C₉H₉IO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00230583 CAS Number: 90347-66-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...