N-Methyl-1-naphthylmethylamine hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H28719-03
H28719-09
AAH28719-03EA
51.6
USD
AAH28719-03
AAH28719-09
N-Methyl-1-naphthylmethylamine hydrochloride 98%
N-Methyl-1-naphthylmethylamine hydrochloride
Purity: 98%
CAS No.: 65473-13-4
Molecular Formula: C12H13N.HCl
Formula Weight: 207.70
Melting Point: 190-193°
MDL No.: MFCD00012555
Formula:
C₁₂H₁₃N Melting Pt: 190…193 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012555 CAS Number: 65473-13-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...