Furan-2,5-dicarboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,5-Furandicarboxylic acid

H28718-06 H28718-03
AAH28718-06EA 254 USD
AAH28718-06 AAH28718-03
Furan-2,5-dicarboxylic acid 98%
Furan-2,5-dicarboxylic acid

Purity: 98% CAS No.: 3238-40-2 Molecular Formula: C6H4O5 Formula Weight: 156.09 Melting Point: 310° MDL No.: MFCD00016582

Formula: C₆H₄O₅
MW: 156.09 g/mol
Melting Pt: >310 °C
Storage Temperature: Ambient
MDL Number: MFCD00016582
CAS Number: 3238-40-2
EINECS: 221-800-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR