3-Aminoisonicotinic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

H28714-03 H28714-06
AAH28714-03EA 176 USD
AAH28714-03 AAH28714-06 AAH28714-MD
3-Aminoisonicotinic acid 95%
3-Aminoisonicotinic acid

Purity: 95% CAS No.: 7579-20-6 Molecular Formula: C6H6N2O2 Formula Weight: 138.13 Melting Point: ca 280° dec. MDL No.: MFCD00137840

Formula: C₆H₆N₂O₂
Melting Pt: ca 280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00137840
CAS Number: 7579-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR