2,4,6-Trivinylboroxin pyridine complex 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Trivinylcyclotriboroxan - Pyridine Complex

H28703-06 H28703-03
AAH28703-06EA 380 USD
AAH28703-06 AAH28703-03
2,4,6-Trivinylboroxin pyridine complex 95%
2,4,6-Trivinylboroxin pyridine complex

Purity: 95% CAS No.: 95010-17-6 Molecular Formula: C6H9B3O3 Formula Weight: 161.57 Storage Temperature: 2°C to 8°C Melting Point: 44-52° MDL No.: MFCD03839940

Formula: C₁₁H₁₄B₃NO₃
MW: 240.67 g/mol
Melting Pt: 44…52 °C
Storage Temperature: Ambient
MDL Number: MFCD03839440
CAS Number: 95010-17-6
EINECS: 672-662-4
UN: 1325
ADR: "4.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR