3-Benzylbenzeneboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
H28682-06
H28682-03
AAH28682-06EA
338
USD
AAH28682-06
AAH28682-03
3-Benzylbenzeneboronic acid pinacol ester 97%
3-Benzylbenzeneboronic acid pinacol ester
Purity: 97%
CAS No.: 1073355-05-1
Molecular Formula: C19H23BO2
Formula Weight: 294.20
Melting Point: 68-69°
MDL No.: MFCD09266179
Formula:
C₁₉H₂₃BO₂ Melting Pt: 68…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD09266179 CAS Number: 1073355-05-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...