4-(Boc-amino)piperidine 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-N-(tert-Butoxycarbonyl)aminopiperidine, tert-Butyl piperidin-4-ylcarbamate, tert-butyl N-(piperidin-4-yl)carbamate, 4-Boc-Aminopiperidine, 4-N-BOC-Aminopiperidine

H28517-03 H28517-06 H28517-14
AAH28517-03EA 51.5 USD
AAH28517-03 AAH28517-06 AAH28517-14
4-(Boc-amino)piperidine 96%
4-(Boc-amino)piperidine

Purity: 96% CAS No.: 73874-95-0 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 Melting Point: 162-166° MDL No.: MFCD00798171

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Melting Pt: 162…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00798171
CAS Number: 73874-95-0
EINECS: 616-026-6
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR