Dimethylphenylsilanol sodium salt 97%

Supplier: Thermo Scientific Chemicals

H28457-03 H28457-06
AAH28457-03EA 116 USD
AAH28457-03 AAH28457-06
Dimethylphenylsilanol sodium salt 97%
Dimethylphenylsilanol sodium salt

Purity: 97% CAS No.: 7646-75-5 Molecular Formula: C8H11NaOSi Formula Weight: 174.25 MDL No.: MFCD09266151

Formula: C₈H₁₁NaOSi
Storage Temperature: Ambient
MDL Number: MFCD09266151
CAS Number: 7646-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR