4-Cyano-4-phenylpiperidine hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

H28414-03 H28414-09
AAH28414-03EA 40.6 USD
AAH28414-03 AAH28414-09
4-Cyano-4-phenylpiperidine hydrochloride 97%
4-Cyano-4-phenylpiperidine hydrochloride

Purity: 97% CAS No.: 51304-58-6 Molecular Formula: C12H14N2.HCl Formula Weight: 222.71 Melting Point: 210-214° UN No.: 3439 MDL No.: MFCD00012775

Formula: C₁₂H₁₅ClN₂
Melting Pt: 210…214 °C
Storage Temperature: Ambient
MDL Number: MFCD00012775
CAS Number: 51304-58-6
EINECS: 257-124-5
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR