L-Phenylalanine benzyl ester hydrochloride 98%
Supplier: Thermo Scientific Chemicals
H28407-06
H28407-14
AAH28407-06EA
90
USD
AAH28407-06
AAH28407-14
L-Phenylalanine benzyl ester hydrochloride 98%
L-Phenylalanine benzyl ester hydrochloride
Purity: 98%
CAS No.: 2462-32-0
Molecular Formula: C16H17NO2.HCl
Formula Weight: 291.77
Storage Temperature: 2°C to 8°C
Melting Point: 197-200°
MDL No.: MFCD00043249
Formula:
C₁₆H₁₇NO₂ MW: 291.78 g/mol Melting Pt: 197…200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043249 CAS Number: 2462-32-0 EINECS: 219-558-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...