4-Chloro-m-anisidine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Amino-2-chloroanisole, 4-Chloro-3-methoxyaniline, 4-Chloro-3-anisidine

H28331-06 H28331-14
AAH28331-03EA 50.09 USD
AAH28331-03 AAH28331-06 AAH28331-14
4-Chloro-m-anisidine 98%
4-Chloro-m-anisidine

Purity: 98% CAS No.: 13726-14-2 Molecular Formula: C7H8ClNO Formula Weight: 157.60 Odor: Characteristic Melting Point: 78-81° UN No.: 2233 MDL No.: MFCD00672967

Formula: C₇H₈ClNO
MW: 157.6 g/mol
Melting Pt: 78…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00672967
CAS Number: 13726-14-2
UN: 2233
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR