Nepsilon-(tert-Butoxycarbonyl)-Nalpha-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Nepsilon-Boc-Nalpha-Fmoc-D-lysine, Fmoc-D-Lys(Boc)-OH
H28301-03
H28301-06
AAH28301-03EA
61.4
USD
AAH28301-03
AAH28301-06
Nepsilon-(tert-Butoxycarbonyl)-Nalpha-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine 98%
Nepsilon-(tert-butoxycarbonyl)-Nalpha-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine
Purity: 98%
CAS No.: 92122-45-7
Molecular Formula: C26H32N2O6
Formula Weight: 468.54
Storage Temperature: 2°C to 8°C
Melting Point: 135-139°
Optical Rotation: 11° (c=1 in DMF)
MDL No.: MFCD00065660
Formula:
C₂₆H₃₂N₂O₆ MW: 468.55 g/mol Melting Pt: 135…139 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065660 CAS Number: 92122-45-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...