3-Chloro-4-methylphenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Chloro-4-methylphenylboronic Acid, 3-Chloro-4-methylbenzeneboronic acid
H28288-03
H28288-06
AAH28288-03EA
23.1
USD
AAH28288-03
AAH28288-06
3-Chloro-4-methylphenylboronic acid 97%
3-Chloro-4-methylphenylboronic acid
Purity: 97%
CAS No.: 175883-63-3
Molecular Formula: C7H8BClO2
Formula Weight: 170.40
Melting Point: 224-230°
MDL No.: MFCD04039010
Formula:
C₇H₈BClO₂ MW: 170.40 g/mol Melting Pt: 224…230 °C Storage Temperature: Ambient |
MDL Number:
MFCD04039010 CAS Number: 175883-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...